Hypersil™ PREP HS Si HPLC Columns
Supplier: Thermo Scientific
41405-152130 41405-252130 41405-054630 41405-104630 41405-154630 41405-254630 41405-059070A 41405-059270A 41405-109070A 41405-109270A 41405-159070A 41405-159270A 41405-159370A 41405-159570A 41405-259070A 41405-259270A 41405-259370A 41405-259570A 41405-018823A 41405-019023A 41405-019323A 41405-029323A 41405-059270XL 41405-109270XL 41405-159270XL 41405-159370XL 41405-159570XL 41405-259270XL 41405-259370XL 41405-259570XL 41405-019323B 41405-029323B 41405-052130 41405-102130
76330-304EA 1129.29 CAD
76330-304 76330-306 76330-308 76330-310 76330-312 76330-314 76330-316 76330-318 76330-320 76330-322 76330-324 76330-326 76330-328 76330-330 76330-332 76330-334 76330-336 76330-338 76330-340 76330-342 76330-346 76330-348 76330-350 76330-352 76330-354 76330-356 76330-358 76330-360 76330-362 76330-382 76330-384 76330-386 76330-494 76330-496
Hypersil™ PREP HS Si HPLC Columns
Chromatography Columns
Maximize retention and loading using Thermo Scientific™ Hypersil™ PREP HS Si preparative HPLC columns.
  • Silica to separate nonpolar and moderately polar organic compounds
  • USP L3 - high surface area particle with 100 Å pore size
  • pH stable from 2 to 7
  • Maximum temperature of 60 °C
Packed with a bare silica, the high surface area particles with a 100 Å pore size are made for normal phase chromatography of nonpolar and moderately polar organic compounds. Our preparative HPLC columns come in a variety of dimensions including an option for XtendedLifeMore Product Information