Eisco LabGlass™ Low Form Glass Beakers, ASTM
Supplier: Wards
CH0124A10PK12 CH0124A20PK12 CH0124A30PK12 CH0124B CH0124C CH0124D CH0124E CH0124F CH0124G CH0124H CH0124I CH0124J
470335-876PK 30.8 CAD
470335-876 470335-878 470335-880 470335-882 470335-884 470335-886 470335-888 470335-890 470335-892 470335-894 470335-896 470335-898
Eisco LabGlass™ Low Form Glass Beakers, ASTM
Beakers
High quality borosilicate 3.3 glass beakers feature a spout for easy pouring as well as double scale graduation markings.
  • Made from high quality borosilicate 3.3 glass
  • Low form design with spout for easy pouring
  • Chemical resistant, double scale white enamel graduations
  • Superior chemical durability
  • Low temperature gradient, can withstand thermal shock
  • Uniform wall thickness for superior durability
  • Autoclavable at 121 °C
  • Graduated per ASTM E960
Eisco ASTM beakers are made of high-quality Borosilicate 3.3 glass. These durable glass beakers have a lowMore Product Information