Anti-PLCB1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-6976R-CY7
10458-904EA 1270.19 CAD
10458-904
Anti-PLCB1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Phospholipase C beta 1 catalyzes the formation of inositol 1,4,5-trisphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. This reaction uses calcium as a cofactor and plays an important role in the intracellular transduction of many extracellular signals. Its gene is activated by two G-protein alpha subunits, alpha-q and alpha-11. Two transcript variants encoding different isoforms have been found for its gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLCB1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR